Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Methionine, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. p-5391643
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Packaging Poly Bottle
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Quantity 100 g
Formula Weight 149.21
Grade Multi-Compendial/USP
Chemical Name or Material L-Methionine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.