Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-(-)-Methioninol, 97%

Click to view available options
Quantity:
250mg
1g
2899-37-8
C5H13NOS
135.225
MFCD00004735
MIQJGZAEWQQAPN-YFKPBYRVSA-N
l-methioninol,l---methioninol,h-methioninol,2s-2-amino-4-methylsulfanyl butan-1-ol,s---methioninol,s-2-amino-4-methylthio butan-1-ol,d-s---methionanol,methioninol,2s-2-amino-4-methylsulfanylbutan-1-ol,s-2-amino-4-methylsulfanyl-butan-1-ol
2724404
(2S)-2-amino-4-methylsulfanylbutan-1-ol
CSCCC(CO)N
2899-37-8
C5H13NOS
135.225
MFCD00004735
MIQJGZAEWQQAPN-YFKPBYRVSA-N
l-methioninol,l---methioninol,h-methioninol,2s-2-amino-4-methylsulfanyl butan-1-ol,s---methioninol,s-2-amino-4-methylthio butan-1-ol,d-s---methionanol,methioninol,2s-2-amino-4-methylsulfanylbutan-1-ol,s-2-amino-4-methylsulfanyl-butan-1-ol
2724404
(2S)-2-amino-4-methylsulfanylbutan-1-ol
CSCCC(CO)N
Melting Point 31°C to 33°C
Quantity 1g
Flash Point >110°C (230°F)
Optical Rotation −21° (c=5 in Water)
Beilstein 2231656
Formula Weight 135.23
Percent Purity 97%
Chemical Name or Material L-(-)-Methioninol