Learn More
Enzo Life Sciences L-NASPA (1mg). CAS: 155915-46-1
SDP
Supplier:  Enzo Life Sciences BMLLP1010001
Encompass_Preferred
L-NASPA is an antagonist of LPA-induced platelet aggregation and chloride conductance in Xenopus oocytes. In human breast carcinoma MDA-MB231 cells it is a potent LPA agonist. Alternative name: N-Palmitoyl-L-serine phosphoric acid. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10 mg/ml)or ethanol (10 mg/ml). Long Term Storage: -80°C.
Catalog No. 50-201-1152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.