Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Norleucine methyl ester hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
3844-54-0
C7H16ClNO2
181.66
MFCD00077151
FMMOVZRXLNVNBI-RGMNGODLSA-N
s-methyl 2-aminohexanoate hydrochloride,h-nle-ome hcl,l-norleucine methyl ester hydrochloride,norleucine, methyl ester, hydrochloride,methyl 2s-2-aminohexanoate hydrochloride,h-nle-ome.hcl,h-nle-ome?cl,h-nle-ome hydrochloride,l-norleucine methyl ester hcl,methyl l-norleucinate hydrochloride
14039065
methyl (2S)-2-aminohexanoate;hydrochloride
CCCCC(C(=O)OC)N.Cl
3844-54-0
C7H16ClNO2
181.66
MFCD00077151
FMMOVZRXLNVNBI-RGMNGODLSA-N
s-methyl 2-aminohexanoate hydrochloride,h-nle-ome hcl,l-norleucine methyl ester hydrochloride,norleucine, methyl ester, hydrochloride,methyl 2s-2-aminohexanoate hydrochloride,h-nle-ome.hcl,h-nle-ome?cl,h-nle-ome hydrochloride,l-norleucine methyl ester hcl,methyl l-norleucinate hydrochloride
14039065
methyl (2S)-2-aminohexanoate;hydrochloride
CCCCC(C(=O)OC)N.Cl
Beilstein 4880001
Formula Weight 181.66
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material L-Norleucine methyl ester hydrochloride