Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Norvaline methyl ester hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
25g
56558-30-6
C6H14ClNO2
167.633
MFCD00190974
IODNYRVZHKRFIW-JEDNCBNOSA-N
s-methyl 2-aminopentanoate hydrochloride,l-norvaline methyl ester hydrochloride,methyl l-norvalinate hcl,h-nva-ome.hcl,h-nva-ome hcl,methyl l-norvalinate hydrochloride,s-2-amino-pentanoic acid methyl ester hydrochloride,s-methyl 2-aminopentanoate,methyl 2s-2-aminopentanoate hydrochloride,h-nva-ome?cl
14118488
methyl (2S)-2-aminopentanoate;hydrochloride
CCCC(C(=O)OC)N.Cl
56558-30-6
C6H14ClNO2
167.633
MFCD00190974
IODNYRVZHKRFIW-JEDNCBNOSA-N
s-methyl 2-aminopentanoate hydrochloride,l-norvaline methyl ester hydrochloride,methyl l-norvalinate hcl,h-nva-ome.hcl,h-nva-ome hcl,methyl l-norvalinate hydrochloride,s-2-amino-pentanoic acid methyl ester hydrochloride,s-methyl 2-aminopentanoate,methyl 2s-2-aminopentanoate hydrochloride,h-nva-ome?cl
14118488
methyl (2S)-2-aminopentanoate;hydrochloride
CCCC(C(=O)OC)N.Cl
Beilstein 3561027
Formula Weight 167.63
Physical Form Solid
Percent Purity 97%
Color White
Quantity 5g
Chemical Name or Material L-Norvaline methyl ester hydrochloride