Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Ornithine methyl ester dihydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
40216-82-8
C6H16Cl2N2O2
219.106
MFCD00190977
SNFZNVWAQDVFAK-XRIGFGBMSA-N
h-orn-ome.2hcl,l-ornithine methyl ester dihydrochloride,methyl l-ornithine dihydrochloride,s-methyl 2,5-diaminopentanoate dihydrochloride,h-orn-ome?hcl,methyl l-ornithine hcl,h-orn-ome 2hcl,h-orn-ome2hcl,l-ornithine, methyl ester, dihydrochloride,h-orn-ome 2 hcl
14273060
methyl (2S)-2,5-diaminopentanoate;dihydrochloride
COC(=O)C(CCCN)N.Cl.Cl
40216-82-8
C6H16Cl2N2O2
219.106
MFCD00190977
SNFZNVWAQDVFAK-XRIGFGBMSA-N
h-orn-ome.2hcl,l-ornithine methyl ester dihydrochloride,methyl l-ornithine dihydrochloride,s-methyl 2,5-diaminopentanoate dihydrochloride,h-orn-ome?hcl,methyl l-ornithine hcl,h-orn-ome 2hcl,h-orn-ome2hcl,l-ornithine, methyl ester, dihydrochloride,h-orn-ome 2 hcl
14273060
methyl (2S)-2,5-diaminopentanoate;dihydrochloride
COC(=O)C(CCCN)N.Cl.Cl
Formula Weight 219.11
Physical Form Powder
Percent Purity 97%
Color White
Quantity 25g
Chemical Name or Material L-Ornithine methyl ester dihydrochloride

EINECSNumber : 254-841-5

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold