Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-(+)-Pantolactone, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.
Solubility
Soluble in water, hardly soluble in alcohol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
L-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.
Solubility
Soluble in water, hardly soluble in alcohol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
Specifications
Melting Point | 89°C to 94°C |
Boiling Point | 120°C to 122°C (15 mmHg) |
Quantity | 250 mg |
Beilstein | 4659425 |
Solubility Information | Soluble in water,hardly soluble in alcohol. |
Optical Rotation | +51° (c=3 in Water) |
Formula Weight | 130.14 |
Percent Purity | 98% |
Chemical Name or Material | L-(+)-Pantolactone |
Safety and Handling
Hazard Category | H318 |
Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
Precautionary Statement | P280-P305+P351+P338-P310 |
TSCA | No |
Recommended Storage | Keep cold |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction