Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(-)-Perillic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
23635-14-5
C10H14O2
166.22
MFCD00062539
CDSMSBUVCWHORP-MRVPVSSYSA-N
s---perillic acid,--perillic acid,lopac-p-7083,4-isopropenylcyclohexene-1-carboxylic acid,s--perillic acid,?-perillic acid,4s-4-isopropenylcyclohexene-1-carboxylic acid
2724160
CHEBI:86939
(4S)-4-prop-1-en-2-ylcyclohexene-1-carboxylic acid
CC(=C)C1CCC(=CC1)C(=O)O
23635-14-5
C10H14O2
166.22
MFCD00062539
CDSMSBUVCWHORP-MRVPVSSYSA-N
s---perillic acid,--perillic acid,lopac-p-7083,4-isopropenylcyclohexene-1-carboxylic acid,s--perillic acid,?-perillic acid,4s-4-isopropenylcyclohexene-1-carboxylic acid
2724160
CHEBI:86939
(4S)-4-prop-1-en-2-ylcyclohexene-1-carboxylic acid
CC(=C)C1CCC(=CC1)C(=O)O
Color Beige to Colorless
Specific Rotation Condition - 102.00 (21.00°C c=2,CH3OH)
Melting Point 129°C to 135°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Rotation -98° to -106° (21°C, 589nm) (c=2, CH3OH)
Assay Percent Range 95%
Beilstein 09, IV, 241
Formula Weight 166.22
Physical Form Crystalline Flakes
Percent Purity ≥94%
Chemical Name or Material L(-)-Perillic acid, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.