Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylalanine methyl ester hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
250g
Packaging:
Glass bottle
7524-50-7
C10H13NO2·HCl
215.68
MFCD00012489
SWVMLNPDTIFDDY-FVGYRXGTSA-N
l-phenylalanine methyl ester hydrochloride,h-phe-ome.hcl,methyl l-phenylalaninate hydrochloride,s-methyl 2-amino-3-phenylpropanoate hydrochloride,unii-47hk4y94ja,h-phe-ome hcl,h-phe-ome hydrochloride,l-phenylalanine, methyl ester, hydrochloride,l-phenylalanine methyl ester hcl
75736
methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
COC(=O)C(CC1=CC=CC=C1)N.Cl
7524-50-7
C10H13NO2·HCl
215.68
MFCD00012489
SWVMLNPDTIFDDY-FVGYRXGTSA-N
l-phenylalanine methyl ester hydrochloride,h-phe-ome.hcl,methyl l-phenylalaninate hydrochloride,s-methyl 2-amino-3-phenylpropanoate hydrochloride,unii-47hk4y94ja,h-phe-ome hcl,h-phe-ome hydrochloride,l-phenylalanine, methyl ester, hydrochloride,l-phenylalanine methyl ester hcl
75736
methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
COC(=O)C(CC1=CC=CC=C1)N.Cl
Melting Point 156.0°C to 160.0°C
Packaging Glass bottle
Quantity 2.5g
Specific Rotation Condition + 37.00 (20.00°C c=2,C2H5OH)
Specific Rotation + 37.00
Infrared Spectrum Authentic
Color White
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula C6H5CH2CH(NH2)CO2CH3·HCl
Beilstein 14,499
Formula Weight 215.68
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material L-Phenylalanine methyl ester hydrochloride, 98%
Show More Show Less

EINECSNumber : 231-383-4