Alfa Aesar™ L-Phenylalanine methyl ester hydrochloride, 99%

Manufacturer: Alfa Aesar™ A1145530

Quantity 250g

 View more versions of this product

Catalog No. AAA1145530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7524-50-7
    Molecular Formula C10H13NO2·HCl
    Formula Weight 215.68
    MDL Number MFCD00012489
    Beilstein 3597948
    Synonym H-Phe-OMe.HCl
    Chemical Name or Material L-Phenylalanine methyl ester hydrochloride

    Specifications

    Formula Weight 215.68
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7524-50-7
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 158° to 162°C
    Optical Rotation +37° (c=2 in ethanol)
    Molecular Formula C10H13NO2·HCl
    Beilstein 3597948
    Synonym H-Phe-OMe.HCl
    Health Hazard 1 Warning
    MDL Number MFCD00012489
    Quantity 250g
    Chemical Name or Material L-Phenylalanine methyl ester hydrochloride
    Assay 99%

    Description

    Applications
    Methyl L-phenylalaninate hydrochloride is used in pharmaceutical, food,animal feed,cosmetic,bio-chemical research etc.

    Solubility
    It is soluble in methanol. (5mg/ ml-clear colorless solution)

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents. Stable under recommended storage conditions. Hygroscopic.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit