Alfa Aesar™ L-Phenylalaninol, 98%

Manufacturer: Alfa Aesar™ A1158603

Quantity 1g

 View more versions of this product

Catalog No. AAA1158603

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 3182-95-4
    Molecular Formula C9H13NO
    Formula Weight 151.21
    MDL Number MFCD00004732
    UN Number UN3259
    Beilstein 2208238
    Synonym (S)-(-)-2-Amino-3-phenyl-1-propanol; H-Phe-ol
    Chemical Name or Material L-Phenylalaninol

    Specifications

    Formula Weight 151.21
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    UN Number UN3259
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 1g
    MDL Number MFCD00004732
    RTECS Number UA6900000
    Optical Rotation −23° (c=5 in ethanol)
    Molecular Formula C9H13NO
    CAS 3182-95-4
    Synonym (S)-(-)-2-Amino-3-phenyl-1-propanol; H-Phe-ol
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    Beilstein 2208238
    Melting Point 89° to 94°C
    Chemical Name or Material L-Phenylalaninol
    Assay 98%

    Description

    Applications
    L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

    Solubility
    Soluble in chloroform, ethyl acetate, ethanol and methanol.

    Notes
    Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit