L-Proline, 99%, Alfa Aesar

Manufacturer: Alfa Aesar A1019922

Quantity 100g

 View more versions of this product

Catalog No. AAA1019922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 147-85-3
    Molecular Formula C5H9NO2
    Formula Weight 115.13
    Chemical Name or Material L-Proline

    Specifications

    Formula Weight 115.13
    Quantity 100g
    Molecular Formula C5H9NO2
    CAS 147-85-3
    Chemical Name or Material L-Proline

    Description

    Applications
    L-Proline is used as asymmetric catalysts in organic synthesis and asymmetric aldol cyclization. It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes. It is a precursor of hydroxyproline in collagen. It is an active component of collagen and involved in the proper functioning of joints and tendons. It finds uses in pharmaceutical, biotechnological applications due to its osmoprotectant property. Further, it is used with ninhydrin in the chromatography.

    Solubility
    Soluble in water and ethanol. Insoluble in ether, butanol and isopropanol.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit