Alfa Aesar™ L-Prolyl diphenylborinate, 95%

Manufacturer: Alfa Aesar™ H5576003

Quantity 1g

 View more versions of this product

Catalog No. AAH5576003

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C17H18BNO2
    Formula Weight 279.15
    MDL Number MFCD22125172
    Synonym Diphenylborinic acid L-prolyl ester
    Chemical Name or Material L-Prolyl diphenylborinate

    Specifications

    Formula Weight 279.15
    Molecular Formula C17H18BNO2
    MDL Number MFCD22125172
    Synonym Diphenylborinic acid L-prolyl ester
    Quantity 1g
    Chemical Name or Material L-Prolyl diphenylborinate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit