Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

l/s-Afadin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA125076

Catalog No. PIPA125076


Add to Cart

Description

Description

Recommended positive controls: MDCK, HepG2, mouse liver, rat brain.

l/s-Afadin Polyclonal antibody specifically detects l/s-Afadin in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

l/s-Afadin
Polyclonal
Unconjugated
Afdn
5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; Mllt4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein AF-6; RP3-431P23.3; RP3-470B24.4; S-afadin
Rabbit
Antigen affinity chromatography
RUO
17356, 26955, 4301, 431695
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
1 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
O35889, P55196, Q9QZQ1
Afdn
Synthetic peptide, conjugated to maleimide-activated BSA, corresponding to residues C-K(1163) L P G D D R L M K N R A D H R S(1179) corresponding to rat I+S Afadin (with n-terminal cysteine).
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit