Learn More
Accuflow SystemsSupplier Diversity Partner Precision Digital Drive Motor With Graphical Interface, Masterflex, L/S, Easy-Load, Ii Pump Head (Thick Wall Tubing), Flow Rates Of 0.001-3400 Ml/Min DFS Item
Manufacturer:  Accuflow SystemsSupplier Diversity Partner 7771252
This precision digital drive and pump head offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever! Flow rates of 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size. Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually. This pump drive includes a 6-ft (1.8m) line cord with IEC 320/CEE22 socket and are shipped with country specific plug/cord set as well as a Masterflex™ L/S™ Easy-Load™ II Pump Head featuring automatic retention which holds high-performance precision (thick-walled) tubing securely in place without manual adjustment. Just load the tubing and start the pump.
Catalog No. NC1475877