Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ L-Selectin/CD62L Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793725PLATES

 View more versions of this product

Catalog No. NB009784


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human L-Selectin/CD62L.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
9.375 pg/mL
6402
RUO
L-Selectin/CD62L
Adaptive Immunity, B Cell Development and Differentiation Markers, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
Human
SELL
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human L-Selectin/CD62L.
Sandwich ELISA
  • Standard: Each vial contains 21 ng of recombinant Human L-Selectin/CD62L.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human L-Selectin/CD62L monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human L-Selectin/CD62L monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    5 Plates
    9.375-600 pg/mL
    CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit