Learn More
L-Selectin/CD62L Mouse anti-Human, Alexa Fluor 594, Clone: 4G8, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9787T100
Catalog No. FAB9787T100
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
L-Selectin/CD62L
4G8
Flow Cytometry 5 uL/10^6 cells
Supernatant
Mouse
IgG1
100 Tests
Primary
6402
Human
Flow Cytometry
Alexa Fluor 594
Supplied in a saline solution containing BSA and Sodium Azide.
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Recombinant human L-Selectin/CD62L Extracellular domain
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Binds to CHO cells expressing the human L-Selectin/CD62L protein. It does not recognize CHO cells expressing either the human E-Selectin or human P-Selectin.