Learn More
L-Selectin/CD62L Mouse anti-Human, Alexa Fluor 647, Clone: 4G8, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9787R100
This item is not returnable.
View return policy
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Human samples. It is validated for Flow Cytometry.Specifications
L-Selectin/CD62L | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Binds to CHO cells expressing the human L-Selectin/CD62L protein. It does not recognize CHO cells expressing either the human E-Selectin or human P-Selectin. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
4G8 | |
Flow Cytometry 5 uL/10^6 cells | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1 | |
Recombinant human L-Selectin/CD62L Extracellular domain | |
100 Tests | |
6402 | |
Human | |
Supernatant |