Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Selectin/CD62L Mouse anti-Human, DyLight™ 550, Clone: DREG56, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142795R

Catalog No. NBP142795R


Add to cart

Description

Description

L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

L-Selectin/CD62L
DREG56
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
SELL
PMA-activated human peripheral blood leukocytes
Protein A purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
Primary
6402.0
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody DREG56 recognizes CD62L/ L-selectin, a 65-76 kDa cell surface protein, expressed by neutrophils, monocytes, and subsets of T, B, and NK cells, that interacts with specific carbohydrates exposed on activated endothelial cells. HLDA V; WS Code S056
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit