Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Selectin/CD62L, Rabbit anti-Human,Bovine,Chimpanzee, Clone: DREG-56, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen L-Selectin/CD62L
Clone DREG-56
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Blocking Assay
Host Species Rabbit
Target Species Human, Bovine, Chimpanzee
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015665
View Documents
Novus Biologicals
NBP2810840.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015666
View Documents
Novus Biologicals
NBP2810840.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Human, Bovine, Chimpanzee Samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Blocking Assay.
Specifications

Specifications

L-Selectin/CD62L
Western Blot, Flow Cytometry, Immunoprecipitation, Blocking Assay
Human, Bovine, Chimpanzee
Primary
DREG-56
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit