Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Serinamide hydrochloride, 95%

Click to view available options
Quantity:
5g
25g
100g
65414-74-6
C3H9ClN2O2
140.567
MFCD00067560
VURWDDZIWBGXCK-DKWTVANSSA-N
l-serinamide hydrochloride,h-ser-nh2.hcl,l-serine amide hydrochloride,s-2-amino-3-hydroxypropanamide hydrochloride,2s-2-amino-3-hydroxypropanamide hydrochloride,propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s,s-2-amino-3-hydroxypropionamide hydrochloride,h-ser-nh hcl,h-ser-nh2cl,h-ser-nh2?cl
16212403
(2S)-2-amino-3-hydroxypropanamide;hydrochloride
C(C(C(=O)N)N)O.Cl
65414-74-6
C3H9ClN2O2
140.567
MFCD00067560
VURWDDZIWBGXCK-DKWTVANSSA-N
l-serinamide hydrochloride,h-ser-nh2.hcl,l-serine amide hydrochloride,s-2-amino-3-hydroxypropanamide hydrochloride,2s-2-amino-3-hydroxypropanamide hydrochloride,propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s,s-2-amino-3-hydroxypropionamide hydrochloride,h-ser-nh hcl,h-ser-nh2cl,h-ser-nh2?cl
16212403
(2S)-2-amino-3-hydroxypropanamide;hydrochloride
C(C(C(=O)N)N)O.Cl
Beilstein 3911773
Formula Weight 140.57
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 185°C to 188°C
Quantity 5g
Chemical Name or Material L-Serinamide hydrochloride