Learn More
Copan Diagnostics L-Shaped Spreaders
SDP
L-shaped spreaders
$198.00 - $515.00
Specifications
Material Plastic
Sterility Sterile
For Use With (Equipment) Culture Plates
Shape L-Shape
For Use With (Application) Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
23-600-897
SDP
View Documents
Copan Diagnostics
174CS01
1/peel pack; 500 spreaders/box
Case of 500 for $354.00
Edge
Add to Cart
23-600-898
SDP
View Documents
Copan Diagnostics
174CS05
5/peel pack; 1000 spreaders/box
Case of 1000 for $515.00
Add to Cart
23-600-899
SDP
View Documents
Copan Diagnostics
174CS10
Cell Spreader
Case of 500 for $198.00
Edge
Add to Cart
Description
Description
L-shaped spreaders
L-Shaped Plastic Spreaders
  • For spreading liquid samples
  • Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
  • Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
  • Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Specifications
Plastic
Culture Plates
Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
Sterile
L-Shape