Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Tartaric acid diammonium salt, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass bottle
3164-29-2
C4H12N2O6
184.148
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+]
3164-29-2
C4H12N2O6
184.148
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+]
Assay Percent Range 99%
Linear Formula [-CH(OH)CO2NH4]2
Chlorine (Cl) 50ppm max.
Merck Index 09, 591
Density 1.6000g/mL
Formula Weight 184.15
Physical Form Crystals
Percent Purity ≥98.5%
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Insoluble in Water 0.005% max.
Iron (Fe) 10ppm max.
Packaging Glass bottle
Specific Rotation +31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
Sulfate 50ppm max.
Sulfated Ash 0.05% max.
Specific Rotation Condition + 32.40 (20.00°C c=1.84,H2O)
pH 6.0 to 7.0 (5% soln.)
Color White
Quantity 5g
Chemical Name or Material L(+)-Tartaric acid diammonium salt, 99%
Show More Show Less

EINECSNumber : 221-618-9

RTECSNumber : WW8050000

TSCA : TSCA