Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric Acid (Fine Granular/NF/FCC), Fisher Chemical

Packaging Poly Pail
Quantity 12kg

 View more versions of this product

Catalog No. A313-12


  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

87-69-4
C4H6O6
150.09
2, 3-Dihydroxybutanedioic Acid
99.7 to 100.5% (NF)
Solid
HOOC(CHOH)2COOH
MFCD00064207
L-(+)-Tartaric Acid
FCC/NF

Specifications

87-69-4
C4H6O6
150.09
2, 3-Dihydroxybutanedioic Acid
Pass Test (FCC)
2mg/kg max. (FCC)
Pass Test (FCC)
0.1% max. (NF)
12.0 to 13.0° (FCC)
<0.1 mbar at 20°C
168°C
White
L-(+)-Tartaric Acid
FCC/NF
Solid
HOOC(CHOH)2COOH
MFCD00064207
0.001% max. (NF)
0.05% max. (FCC)
0.5% max. (FCC)
Poly Pail
Meets Requirements (NF)
Pass Test (FCC)
Tartaric Acid
1.6
12kg
99.7 to 100.5% (NF)

Safety and Handling

SDS & Safety Documents

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit