Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Thiocitrulline dihydrochloride, 98%, Thermo Scientific™

Catalog Number AC350290100
Click to view available options
Quantity:
10 mg
212051-53-1
C6H15Cl2N3O2S
264.165
MFCD00270911
JZZQYCYVKMVMJU-FHNDMYTFSA-N
l-thiocitrulline, dihydrochloride, 2-thioureido-l-norvaline, l-thiocitrulline dihydrochloride, s-2-amino-5-thioureidopentanoic acid dihydrochloride, 2s-2-amino-5-carbamothioylamino pentanoic acid dihydrochloride, c6h13n3o2s.2hcl
2734085
(2S)-2-amino-5-(carbamothioylamino)pentanoic acid;dihydrochloride
C(CC(C(=O)O)N)CNC(=S)N.Cl.Cl
212051-53-1
C6H15Cl2N3O2S
264.165
MFCD00270911
JZZQYCYVKMVMJU-FHNDMYTFSA-N
l-thiocitrulline, dihydrochloride, 2-thioureido-l-norvaline, l-thiocitrulline dihydrochloride, s-2-amino-5-thioureidopentanoic acid dihydrochloride, 2s-2-amino-5-carbamothioylamino pentanoic acid dihydrochloride, c6h13n3o2s.2hcl
2734085
(2S)-2-amino-5-(carbamothioylamino)pentanoic acid;dihydrochloride
C(CC(C(=O)O)N)CNC(=S)N.Cl.Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 65°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 10 mg
Formula Weight 264.18
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material L-Thiocitrulline dihydrochloride, 98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit