Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Threonine, ≥98.5%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
72-19-5
C4H9NO3
119.12
MFCD00064270
AYFVYJQAPQTCCC-GBXIJSLDSA-N
l-threonine,threonine,threonin,h-thr-oh,2s,3r-2-amino-3-hydroxybutanoic acid,s-threonine,l---threonine,2-amino-3-hydroxybutyric acid,threonine van,threoninum latin
6288
CHEBI:16857
(2S,3R)-2-amino-3-hydroxybutanoic acid
CC(C(C(=O)O)N)O
72-19-5
C4H9NO3
119.12
MFCD00064270
AYFVYJQAPQTCCC-GBXIJSLDSA-N
l-threonine,threonine,threonin,h-thr-oh,2s,3r-2-amino-3-hydroxybutanoic acid,s-threonine,l---threonine,2-amino-3-hydroxybutyric acid,threonine van,threoninum latin
6288
CHEBI:16857
(2S,3R)-2-amino-3-hydroxybutanoic acid
CC(C(C(=O)O)N)O
Linear Formula CH3CH(OH)CH(NH2)COOH
Packaging Plastic bottle
Formula Weight 119.12
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Melting Point 255°C (decomposition)
Quantity 1kg
Chemical Name or Material L-Threonine