missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, 98.5 to 101.5% (Dry Basis), Affymetrix/USB™

missing translation for 'orderingAttributeHoverText'
Quantity:
100g
1kg
Packaging:
Plastic bottle
73-22-3
C11H12N2O2
204.229
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
l-tryptophan,tryptophan,l-tryptophane,s-tryptophan,tryptophane,h-trp-oh,optimax,trofan,tryptacin,ardeytropin
6305
CHEBI:16828
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
73-22-3
C11H12N2O2
204.229
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
l-tryptophan,tryptophan,l-tryptophane,s-tryptophan,tryptophane,h-trp-oh,optimax,trofan,tryptacin,ardeytropin
6305
CHEBI:16828
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
Packaging Plastic bottle
Formula Weight 204.23
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Melting Point 280°C to 285°C
Quantity 1kg
Chemical Name or Material L-Tryptophan
Assay Dry basis