Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Tryptophan ethyl ester hydrochloride, 97%

Click to view available options
Quantity:
25g
100g
500g
2899-28-7
C13H17ClN2O2
268.741
MFCD00038993
PESYCVVSLYSXAK-MERQFXBCSA-N
l-tryptophan ethyl ester hydrochloride,ethyl l-tryptophanate hydrochloride,h-trp-oet.hcl,s-tryptophan ethyl ester hydrochloride,ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride,h-trp-oet hcl,ethyl l-tryptophanate hcl,ethyl l-tryptophanate monohydrochloride,h-trp-oet. hcl,pubchem10920
134519
ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
CCOC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
2899-28-7
C13H17ClN2O2
268.741
MFCD00038993
PESYCVVSLYSXAK-MERQFXBCSA-N
l-tryptophan ethyl ester hydrochloride,ethyl l-tryptophanate hydrochloride,h-trp-oet.hcl,s-tryptophan ethyl ester hydrochloride,ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride,h-trp-oet hcl,ethyl l-tryptophanate hcl,ethyl l-tryptophanate monohydrochloride,h-trp-oet. hcl,pubchem10920
134519
ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
CCOC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
Beilstein 3919010
Formula Weight 268.74
Physical Form Powder
Percent Purity 97%
Color Brown
Melting Point 220°C to 225°C
Quantity 500g
Chemical Name or Material L-Tryptophan ethyl ester hydrochloride

EINECSNumber : 220-786-0

TSCA : No

Recommended Storage : Ambient temperatures