Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine, +99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
2.5kg
Packaging:
Glass Bottle
Plastic bottle
60-18-4
C9H11NO3
181.191
MFCD00002606
OUYCCCASQSFEME-QMMMGPOBSA-N
l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
6057
CHEBI:17895
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)O
60-18-4
C9H11NO3
181.191
MFCD00002606
OUYCCCASQSFEME-QMMMGPOBSA-N
l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l
6057
CHEBI:17895
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)O
Loss on Drying 0.5% max. (1 g, 105°C)
Specific Rotation Condition − 11.05 (20.00°C c=5,dilHCL/H2O 50/50)
Packaging Plastic bottle
Sulfated Ash 0.1% max.
Color White
Quantity 2.5kg
Specific Rotation -9.8° to -12.3° (20°C, 589nm) (c=5, 1N HCl)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Beilstein 14, 605
Merck Index 15, 10020
Formula Weight 181.19
Physical Form Crystalline Powder
Percent Purity ≥99.0% (on dried substance)
Chemical Name or Material L-Tyrosine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.