Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Tyrosine benzyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
52799-86-7
MFCD00191034
(S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl
52799-86-7
MFCD00191034
(S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl
Beilstein 2901434
Formula Weight 271.32
Physical Form Solid
Percent Purity 95%
Melting Point 94°C to 96°C
Quantity 25g
Chemical Name or Material L-Tyrosine benzyl ester