Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine Disodium Salt, Dihydrate, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
122666-87-9
C9H15NNa2O5
263.20
FBKIASNRVHFWNA-UHFFFAOYNA-N
sodium s-2-amino-3-4-hydroxyphenyl propanoate
66602062
2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium
O.O.[Na].[Na].NC(CC1=CC=C(O)C=C1)C(O)=O
122666-87-9
C9H15NNa2O5
263.20
FBKIASNRVHFWNA-UHFFFAOYNA-N
sodium s-2-amino-3-4-hydroxyphenyl propanoate
66602062
2-amino-3-(4-hydroxyphenyl)propanoic acid dihydrate disodium
O.O.[Na].[Na].NC(CC1=CC=C(O)C=C1)C(O)=O

High quality biochemicals for research use

Packaging Poly Bottle
Formula Weight 261.19
Grade BAKER ANALYZED™ Biochemical Reagent
Quantity 100g
Chemical Name or Material L-Tyrosine Disodium Salt, Dihydrate