Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Tyrosine ethyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
949-67-7
C11H15NO3
209.245
MFCD00063046
SBBWEQLNKVHYCX-JTQLQIEISA-N
l-tyrosine ethyl ester,ethyl l-tyrosinate,tyrosine ethyl ester,h-tyr-oet,l-tyrosine, ethyl ester,tyr-oet,ethyl tyrosine ester,l-tyrosine ethyl,unii-17rpw76a9g,s-ethyl 2-amino-3-4-hydroxyphenyl propanoate
70364
CHEBI:9798
ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CCOC(=O)C(CC1=CC=C(C=C1)O)N
949-67-7
C11H15NO3
209.245
MFCD00063046
SBBWEQLNKVHYCX-JTQLQIEISA-N
l-tyrosine ethyl ester,ethyl l-tyrosinate,tyrosine ethyl ester,h-tyr-oet,l-tyrosine, ethyl ester,tyr-oet,ethyl tyrosine ester,l-tyrosine ethyl,unii-17rpw76a9g,s-ethyl 2-amino-3-4-hydroxyphenyl propanoate
70364
CHEBI:9798
ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CCOC(=O)C(CC1=CC=C(C=C1)O)N
Beilstein 2215181
Formula Weight 209.25
Physical Form Solid
Percent Purity 95%
Melting Point 103°C to 107°C
Quantity 5g
Chemical Name or Material L-Tyrosine ethyl ester

TSCA : TSCA

storageNote1 : -30° to -10°C