Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Tyrosine methyl ester hydrochloride, 98+%

Click to view available options
Quantity:
25g
100g
3417-91-2
C10H14ClNO3
231.676
MFCD00012607
VXYFARNRGZWHTJ-FVGYRXGTSA-N
l-tyrosine methyl ester hydrochloride,h-tyr-ome.hcl,methyl l-tyrosinate hydrochloride,h-tyr-ome hcl,methyl tyrosinate hydrochloride,unii-w42m0mi271,tyrosine methyl ester hydrochloride,methyl l-tyrosinate hcl,s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride,s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride
76956
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC=C(C=C1)O)N.Cl
3417-91-2
C10H14ClNO3
231.676
MFCD00012607
VXYFARNRGZWHTJ-FVGYRXGTSA-N
l-tyrosine methyl ester hydrochloride,h-tyr-ome.hcl,methyl l-tyrosinate hydrochloride,h-tyr-ome hcl,methyl tyrosinate hydrochloride,unii-w42m0mi271,tyrosine methyl ester hydrochloride,methyl l-tyrosinate hcl,s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride,s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride
76956
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
COC(=O)C(CC1=CC=C(C=C1)O)N.Cl
Beilstein 3917353
Formula Weight 231.68
Percent Purity ≥98%
Melting Point 187°C (decomposition)
Quantity 25g
Optical Rotation −5.2° (c= 2.4 in Water)
Chemical Name or Material L-Tyrosine methyl ester hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C