Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine tert-butyl ester, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16874-12-7
C13H19NO3
237.299
DIGHFXIWRPMGSA-NSHDSACASA-N
l-tyrosine tert-butyl ester,h-tyr-otbu,tert-butyl l-tyrosinate,l-tyrosine t-butyl ester,tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate,h-tyr-obut,tyrosine, 1,1-dimethylethyl ester,2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester,l-tyrosine tert.butyl ester,pubchem7068
6950582
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
16874-12-7
C13H19NO3
237.299
DIGHFXIWRPMGSA-NSHDSACASA-N
l-tyrosine tert-butyl ester,h-tyr-otbu,tert-butyl l-tyrosinate,l-tyrosine t-butyl ester,tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate,h-tyr-obut,tyrosine, 1,1-dimethylethyl ester,2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester,l-tyrosine tert.butyl ester,pubchem7068
6950582
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
Quantity 5g
Specific Rotation 25
Packaging Glass bottle
Melting Point 142.5°C to 143.8°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 237.3
Physical Form Powder
Percent Purity 99%
Chemical Name or Material L-Tyrosine tert-butyl ester
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.