Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Tyrosylglycine, 95%

Click to view available options
Quantity:
250mg
1g
673-08-5
C11H14N2O4
238.243
MFCD00037779
HPYDSVWYXXKHRD-VIFPVBQESA-N
h-tyr-gly-oh,tyrosyl-glycine,tyr-gly,chembl89956,s-2-2-amino-3-4-hydroxyphenyl propanamido acetic acid,tyrosylglycine,l-tyrosylglycine,2s-2-amino-3-4-hydroxyphenyl propanamido acetic acid,glycine, l-tyrosyl,l-tyrosyl-glycine
7021854
CHEBI:74990
2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]acetic acid
C1=CC(=CC=C1CC(C(=O)NCC(=O)O)N)O
673-08-5
C11H14N2O4
238.243
MFCD00037779
HPYDSVWYXXKHRD-VIFPVBQESA-N
h-tyr-gly-oh,tyrosyl-glycine,tyr-gly,chembl89956,s-2-2-amino-3-4-hydroxyphenyl propanamido acetic acid,tyrosylglycine,l-tyrosylglycine,2s-2-amino-3-4-hydroxyphenyl propanamido acetic acid,glycine, l-tyrosyl,l-tyrosyl-glycine
7021854
CHEBI:74990
2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]acetic acid
C1=CC(=CC=C1CC(C(=O)NCC(=O)O)N)O
Beilstein 2142807
Formula Weight 238.24
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material L-Tyrosylglycine

storageNote1 : 2° to 8°C