Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Valine, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

72-18-4
C5H11NO2
117.148
KZSNJWFQEVHDMF-BYPYZUCNSA-N
6287
CHEBI:16414
CAS 72-18-4
Molecular Formula C5H11NO2
Packaging Poly Bottle
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILES CC(C)C(C(=O)O)N
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
Molecular Weight (g/mol) 117.148
PubChem CID 6287
ChEBI CHEBI:16414
Formula Weight 117.15
Quantity 100g
Specific Gravity 1L = 1.23kg
Chemical Name or Material L-Valine
Grade Multi-Compendial/USP
Show More Show Less