Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (Native and denatured), Thermo Scientific™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$170.00 - $946.00

Specifications

Antigen L1 Cell Adhesion Molecule Ab-1
Clone UJ127
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS770B
View Documents
Thermo Scientific™
MS770B
Biotinylated Ab with BSA and Azide 1mL
Each for $946.00
Add to cart
 
MS770B0
View Documents
Thermo Scientific™
MS770B0
Biotinylated Ab with BSA and Azide 100μL
Each for $231.00
Add to cart
 
MS770B1
View Documents
Thermo Scientific™
MS770B1
Biotinylated Ab with BSA and Azide 500μL
Each for $627.50
Add to cart
 
MS770P
View Documents
Thermo Scientific™
MS770P
Purified Ab with BSA and Azide 1mL
Each for $743.50
Add to cart
 
MS770P0
View Documents
Thermo Scientific™
MS770P0
Purified Ab with BSA and Azide 100μL
Each for $170.00
Add to cart
 
MS770P1
View Documents
Thermo Scientific™
MS770P1
Purified Ab with BSA and Azide 500μL
Each for $497.00
Add to cart
 
MS770P1ABX
View Documents
Thermo Scientific™
MS770P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $536.00
Add to cart
 
MS770PABX
View Documents
Thermo Scientific™
MS770PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $788.00
Add to cart
 
MS770R7
View Documents
Thermo Scientific™
MS770R7
Ready-to-use for IHC (UVLP) 7mL
Each for $519.50
Add to cart
 
MS770R1
View Documents
Thermo Scientific™
MS770R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (Native and denatured), Thermo Scientific™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, adductus thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, neural migration and in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising from neuroectoderm.

Host Species: Mouse

Clone: UJ127

Isotype: IgG1/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Homogenous suspension of 16-week human fetal brain

Molecular Weight: 220-240kDa

Positive Control: IMR-5 cells Neuroblastoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

L1 Cell Adhesion Molecule Ab-1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cancer and Tumor Biology
Homogenous suspension of 16 week human fetal brain
UJ127
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit