Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Mouse anti-Human, Mouse, Rat, APC, Clone: OTI2A6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271097APC
Description
L1CAM Monoclonal specifically detects L1CAM in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
L1CAM | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
L1CAM | |
Full length recombinant protein of human L1CAM (NP_005521) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI2A6 | |
Flow Cytometry | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Mouse | |
Affinity Purified | |
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
3897.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title