Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Mouse anti-Human, Clone: UJ127, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
$206.00 - $493.00
Specifications
Antigen | L1CAM |
---|---|
Clone | UJ127.11 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 - 5 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug/mL, Immunofluorescence 1 - 2 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002682SS
|
Novus Biologicals
NB1002682SS |
0.025 mg |
Each for $206.00
|
|
NB1002682
|
Novus Biologicals
NB1002682 |
0.1 mg |
Each for $493.00
|
|
Description
L1CAM Monoclonal specifically detects L1CAM in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Immunofluorescence, CyTOF-ready.Specifications
L1CAM | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
Unconjugated | |
Mouse | |
Cellular Markers, Glia Markers, Neuroscience | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
L1CAM | |
IgG1 κ | |
Protein A or G purified |
UJ127.11 | |
Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 - 5 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug/mL, Immunofluorescence 1 - 2 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
3897 | |
Homogenous suspension of 16 week human foetal brain. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title