Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1CAM Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen L1CAM
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093953
View Documents
Novus Biologicals
NBP303276100UL
100 μg
Each for $320.71
Add to cart
 
NB093954
View Documents
Novus Biologicals
NBP30327620UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

L1CAM Polyclonal antibody specifically detects L1CAM in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

L1CAM
Unconjugated
Rabbit
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
PBS with 50% glycerol, pH7.3.
3897
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Recombinant fusion protein containing a sequence corresponding to amino acids 1143-1257 of human L1CAM (NP_000416.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit