Learn More
L1CAM Rat anti-Mouse, Alexa Fluor 488, Clone: 555, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB5674G100UG
Catalog No. FAB5674G100
Add to cart
This item is not returnable. View return policy
Description
Description
L1CAM Monoclonal specifically detects L1CAM in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
L1CAM
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Rat
IgG2a
RUO
Primary
3897
Mouse
Flow Cytometry
555
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
L1CAM
Mouse cerebellum-derived partially purified L1CAM
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse L1CAM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEPCAM, rhMCAM, rhNCAM, rhNCAM-L1, rhOBCAM, recombinant mouse (rm)MAdCAM-1, or rmOCAM is observed.
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)