Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Rat anti-Mouse, Alexa Fluor 594, Clone: 555, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB5674T100UG
This item is not returnable.
View return policy
Description
L1CAM Monoclonal specifically detects L1CAM in Mouse samples. It is validated for Flow Cytometry.Specifications
L1CAM | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse L1CAM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEPCAM, rhMCAM, rhNCAM, rhNCAM-L1, rhOBCAM, recombinant mouse (rm)MAdCAM-1, or rmOCAM is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
555 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
L1CAM | |
Mouse cerebellum-derived partially purified L1CAM | |
RUO | |
3897 | |
Mouse | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)