Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ L3MBTL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583723
Description
Immunogen sequence: LSLKADTKED GEERDDEMEN KQDVRILRGS QRARRKRRGD SAVLKQGLPP KGKKAWCWAS YLEEEKAVAV PAKLFKEH.
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
Specifications
L3MBTL3 | |
Polyclonal | |
Unconjugated | |
L3MBTL3 | |
AI481284; H-l(3)mbt-like protein 3; KIAA1798; l(3)mbt-like 3; l(3)mbt-like 3 (Drosophila); l(3)mbt-like protein 3; L3MBTL histone methyl-lysine binding protein 3; L3mbtl3; L3MBTL3, histone methyl-lysine binding protein; lethal(3)malignant brain tumor-like protein 3; lethal(3)malignant brain tumor-like protein 3; LOW QUALITY PROTEIN: lethal(3)malignant brain tumor-like protein 3; MBT1; MBT-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84456 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JM7 | |
L3MBTL3 | |
Recombinant protein corresponding to Human L3MBTL3. Recombinant protein control fragment (Product #RP-106066). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction