Learn More
Lab-Aids AIDS and STD Transmission and Control Kit
Simulate disease transmission and methods of prevention and study effectiveness of latex barriers in the transmission of disease.
$129.00 - $246.00
Specifications
Class Size | 30 students |
---|---|
For Use With (Application) | Teaches and informs students by simulating the transmission of sexual diseases |
Description
This very relevant hands-on activity illustrates how easily sexually transmitted diseases such as Hepatitis B and AIDS can spread through teenage populations. Students test results of their various exchanges. They are able to identify the contact that infected them or determine if they are perhaps the first case or 'Patient Zero'. Students also explore the permeability and effectiveness of latex barriers in the prevention of sexually transmitted diseases.
Refill kit includes:
- 30 bottles, labeled “body fluids” no. 1 to 30
- 3 drop control bottles of “aids testing solution”
- 3 drop control bottles of “hepatitis b testing solution”
- 1 vial of fragrance 1 (lemon)
- 1 vial of fragrance 2 (vanilla)
- 1 vial of fragrance 3 (almond)
- 1 Vial of Fragrance 4 (peppermint)
- 1 Vial of Fragrance 5 (banana)
- 1 Vial of Fragrance 6 (control)
- 1 Balloons, latex, 144/pkg
- 1 Petroleum jelly, tube
Specifications
30 students | |
Teaches and informs students by simulating the transmission of sexual diseases |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.