Learn More
Lab-Aids™ Contaminants and the Water Cycle
SCI_ED
In this modeling activity, students follow a molecule of water as it travels through the water cycle.
Manufacturer:  Lab-Aids™ 434S
Catalog No. S98813
Add to cart
Includes: 12 sets of six Water Cycle cards; transparency: “Sample Responses to Analysis Question 1”; 12 white number cubes; 12 blue number cubes; teacher's guide; and 28 student worksheets and guides
Description
Description
The Contaminants and the Water Cycle activity kit can be used to model both the basic water cycle and the water cycle with biological and chemical contaminants that are picked up along the way. Depending upon your situation, students can do the activity once, with or without contaminants, or they can do it twice, initially without contaminants and then later with contaminants. Students start at some point where water exits on Earth, such as in a river or a raindrop. Students then roll a dice and consult Water Cycle Cards to determine where their water molecule will go next. After making at least six rolls of the dice, each student writes a story that demonstrates an understanding of their water molecules travels within the water cycle (and describes the types and sources of contaminants picked up along the way).
Specifications
Specifications
12 Sets of six water cycle cards, transparency “sample responses to analysis question 1”, 12 white number cubes, 12 blue number cubes, teacher's guide, and 28 student worksheets and guides
Contaminants and the Water Cycle Kit
Used to model both the basic water cycle and the water cycle with biological and chemical contaminants that are picked up along the way