Learn More
Lab-Aids™ DNA Fingerprinting Diagnostics (Replication, PCR and RFLP) Products for Science Education
Study how DNA analysis is used to diagnose disease. Lab-Aids™ DNA Fingerprinting Diagnostics (Replication, PCR and RFLP) introduces biotechnology, forensic science, or AP Biology students to the concepts of DNA replication and how it relates to PCR and RFLP analyses of DNA
Manufacturer:  Lab-Aids™ 75
Catalog No. S11175
Add to cart
Includes: 1 teachers guide; 20 student worksheets and guides; 10 nucleotides sheet: cytosine 28 (blue); 10 nucletoides sheet: guanine 28 (yellow); 10 nucleotides sheet: adenine 66 (red); 10 nucleotides sheet: thymine 66 (green); 10 DNA stands, labels, sheets; 2 mutant CFTR; 2 normal CFTR; 2 primer 1; 2 primer 2; 2 template-mutant CFTR; 2 template-normal CFTR; 10 electrophoresis bands, labels, sheets; 1 mutant CFTR PCR product (22 bp); 1 mutant CFTR RFLP product (10 bp); 1 mutant CFTR RFLP product (14 bp); 1 normal CFTR PCR product (25 bp); 1 normal CFTR RFLP product (25 bp); 60 adhesive strips sized for 4 bp, 30 18 bp, 30 21 bp; 30 adhesive strips sized for 22 bp,3 25 bp; 10 DNA polymerase; 3 glue sticks, removable; 10 bags, resealable; 10 taql restriction enzyme; 10 workboards, two-sided
Description
Description
In addition to studying the concepts of DNA replication and how it relates to PCR (Polymerase Chain Reaction) and RFLP (Restriction Fragment Length Polymorphism) analyses of DNA, students also observe how DNA analyses can be used to diagnose disease. This activity is a great introduction or review if you have the equipment and materials to do gel electrophoresis and DNA fingerprinting
  • Materials for 30 students
  • Reusable
  • Requires one 50 minute class period
  • Includes teachers guide and student worksheets with literacy activities
Concepts Illustrated:
  • Construct genes with the nucleotide configuration for cystic fibrosis
  • Observe differences between normal and mutant genes
  • Discover how disease can be diagnosed by using two methods of DNA fingerprinting
  • Demonstrate DNA amplification and PCR
  • Simulate gel electrophoresis
Specifications
Specifications
Dna Fingerprinting Diagnostics (Replication, Pcr And Rflp) Kit (Developed by Lab-Aids™)
30 students
1 teachers guide; 20 student worksheets and guides; 10 nucleotides sheet: cytosine 28 (blue); 10 nucletoides sheet: guanine 28 (yellow); 10 nucleotides sheet: adenine 66 (red); 10 nucleotides sheet: thymine 66 (green); 10 DNA stands, labels, sheets; 2 mutant CFTR; 2 normal CFTR; 2 primer 1; 2 primer 2; 2 template-mutant CFTR; 2 template-normal CFTR; 10 electrophoresis bands, labels, sheets; 1 mutant CFTR PCR product (22 bp); 1 mutant CFTR RFLP product (10 bp); 1 mutant CFTR RFLP product (14 bp); 1 normal CFTR PCR product (25 bp); 1 normal CFTR RFLP product (25 bp); 60 adhesive strips sized for 4 bp, 30 18 bp, 30 21 bp; 30 adhesive strips sized for 22 bp,3 25 bp; 10 DNA polymerase; 3 glue sticks, removable; 10 bags, resealable; 10 taql restriction enzyme; 10 workboards, two-sided