Learn More
Lab-Aids™ DNA Staining Kit Products for Science Education
Macerate, stain and mount selected root tips, locate and diagram DNA in cells and observe and draw various stages of cell division.
Manufacturer:  Lab-Aids™ 170
Catalog No. S19394F
Add to cart
Includes:
  • Worksheets
  • Teacher's guide
Description
Description
This highly engaging kit is based on the Feulgen reaction where DNA in plant cells are stained purple for easy viewing. Students macerate, stain, and mount specially grown root tips (provided in the kit). Then, they locate and diagram DNA in the cells. Finally, the students are asked to observe and draw the various stages of cell division. This enables the students to develop a better understanding of the hereditary material in plant chromosomes.
All aspects of the lab activity are performed by the individual student. Microscopes are needed.
Accommodates 2 classes, each with 15 groups of 2 students.
Order Info
Class size: 30 students.
Specifications
Specifications
30 students
Teacher's guide with MSDS, 30 student worksheets and guides, 30 microscope slides, 30 cover slips, 30 Plastic treating dishes, 30 transfer brushes, specially selected root tips, vial, dilute Acetic Acid, drop control bottle, Hydrochloric Acid, 1 M, drop control bottle, 2 Leucofuchsin dye, 2 drop control bottle, 2 Sodium Bisulfite solution, drop control bottle
Microscope
Education
For easy viewing dna
Dna Staining Kit
50 minutes