Learn More
Lab-Aids™ Investigating and Applying Genetics Kit Products for Science Education
Explore selective breeding for desired traits in crops such as corn.
Manufacturer:  Lab-Aids™ 603S
Catalog No. S90181
Add to cart
Includes:
  • 64 Lab-Aids corn ears
  • Two color transparencies
  • 32 student activity worksheets and guides
  • 32 readings on breeding crops
  • Instruction manual
Description
Description
Investigate the crosses of hybrid and dihybrid plants for two unlinked genetic traits — corn kernel color and sweetness.
  • Predict and analyze the ratio of phenotypes produced by different crosses
  • Discuss the differences between traditional selective breeding and modern genetic engineering
  • Understand the pattern and mechanism of inheritance for Mendelian traits
  • Class size: 32 students
Specifications
Specifications
Investigating And Applying Genetics Kit
32 students
Teacher's guide, 32 student worksheets and guides - part 1 and 2, 32 breeding crops with desirable traits reading, 4 color transparency, 16 each corn “ear a, b, c, d”, lab-aids™
50 minutes
For desired traits in crops such as corn