Learn More
Lab-Aids™ Investigating Hard and Soft Water Kit Products for Science Education
Gain experience with ionization, precipitation and solubility. Applies to ecology, earth science and consumer education.
$57.50 - $83.00
Includes:
  • 36 test tubes
  • 12 plastic spatulas
  • Bottle of soap chips
  • Two bottles of Deionized water
  • 2 bottles of Hard water
  • Bottle of Calcium chloride solution
  • Bottle of Magnesium chloride solution
  • 2 bottles of soap solution
  • 2 vials of sodium carbonate
  • 50 student worksheets and guides
  • Instruction manual with MSDS
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S94511 Products for Science Education
View Documents
Lab-Aids™
191
Investigating Hard and Soft Water Kit
Each for $83.00
Add to cart
S94513 Products for Science Education
View Documents
Lab-Aids™
191RC
Refill for Investigating Hard and Soft Water Kit
Each for $57.50
Add to cart
Description
Description
Using simple tests, students gain experience with ionization, precipitation and solubility. This Lab-Aid™ kit is an excellent activity for studying water purification, water pollution, ecology, earth science and consumer education. Students study important concepts in a clear, meaningful and relevant manner. This kit is highly recommended for intermediate and junior high science courses. Accommodates 2 classes, each with 12 groups of 2 students. Includes Teacher guide, student sheets, assessment questions and SDS for all chemicals.
Refill Kit Includes:
  • 1 Drop control bottle of Soap Material
  • 2 Bottles of Deionized Water
  • 2 Bottles of Hard water
  • 1 Drop control bottle of Calcium Chloride solution
  • 1 Drop control bottle of Magnesium Chloride Solution
  • 2 Drop control bottle of Soap Solution
  • 2 Vials of Calcium Sulfate
  • 1 Vial of Sodium Carbonate
Order Info
Highly recommended for intermediate and junior high science courses.