Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab-Aids™ Normal Mitosis Kit
SCI_ED

Prepare a squashed stained slide of a root tip, observe cells undergoing mitosis and gain experience in microtechnique and staining.

Manufacturer:  Lab-Aids™ 9

Catalog No. S19354


Add to cart

Includes:
  • Root tips
  • Plastic tissue-treating dishes
  • Microscope slides and cover slips
  • Four bottles of solution A (alcohol and hydrochloric acid)
  • Four bottles of solution B (acetic acid, chloroform, alcohol)
  • Tissue-macerating blades
  • Four vials aceto-orcein stain
  • Tissue transfer brushes
  • Student worksheets and guides

Requires : Microscope

Description

Description

Using this kit, students will be challenged as they identify the phases of plant mitosis, observe chromosomal development, gain hands-on experience with the micro-techniques, stain and mount their own slides. Students prepare their own plant cells by treating scientifically grown onion root tips cells with a mild acid, then staining the cells. A stained section of cells is then squashed onto a slide and smeared into a single layer for microscopic study. All of the activities in this kit can be completed in a standard lab period. Kit is for 30 students.

Order Info

Class size: 30 students.

Specifications

Specifications

Helps in identify the phases of plant mitosis, observe chromosomal development, gain hands-on experience with the micro-techniques, stain and mount their own slides
Normal Mitosis Kit
30 Students
Teacher's guide with MSDS, 30 student worksheets and guides, 30 microscopic slides, 30 cover slips, 30 tissue transfer brushes, 30 tissue treating trays with cavities A and B, root tips scientifically grown and fixed vial, 4 drop control bottles of solution A and B, 4 drop control bottles of aceto orcein stain
One 50 minute class period
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit